Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tspan11
Entrez Gene ID
312727
Gene Name
tetraspanin 11
Gene Aliases
RGD1305424
Chromosome Location
Chr.4: 160455848 - 160522905 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024262.1 NP_001019433.1 5 - 6 584 67
GenBank mRNA BC092652.1 - 5 - 6 584 67

View Full Product Details