Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tspan11
Entrez Gene ID
312727
Gene Name
tetraspanin 11
Gene Aliases
RGD1305424
Chromosome Location
Chr.4: 160455848 - 160522905 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024262.1 NP_001019433.1 6 - 7 746 96
XM_008763291.1 XP_008761513.1 7 - 8 952 96
GenBank mRNA BC092652.1 - 6 - 7 746 96

View Full Product Details