Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B4galnt3
Entrez Gene ID
500306
Gene Name
beta-1,4-N-acetyl-galactosaminyl transferase 3
Gene Aliases
RGD1561056
Chromosome Location
Chr.4: 152736060 - 152835488 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191976.1 NP_001178905.1 19 - 20 2864 134
XM_006237267.2 XP_006237329.1 19 - 20 3167 134

View Full Product Details