The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017099.4 | NP_058795.3 | 2 - 3 | 555 | 66 |
GenBank mRNA | AB043636.1 | - | 1 - 2 | 385 | 66 |
AB043637.1 | - | 1 - 2 | 385 | 66 | |
BC078863.1 | - | - | 538 | 66 | |
D42145.1 | - | 2 - 3 | 558 | 66 |