Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scnn1a
Entrez Gene ID
25122
Gene Name
sodium channel, non-voltage-gated 1 alpha subunit
Gene Aliases
-
Chromosome Location
Chr.4: 157835474 - 157860513 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031548.2 NP_113736.1 7 - 8 1805 66
GenBank mRNA BC070902.1 - 7 - 8 1805 66
EU627515.1 - 7 - 8 1440 66
U54699.1 - 7 - 8 1472 66
U54700.1 - 7 - 8 1472 66
X70497.1 - 7 - 8 1518 66
X70521.1 - 7 - 8 1493 66

View Full Product Details