Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mynn
Entrez Gene ID
361924
Gene Name
myoneurin
Gene Aliases
-
Chromosome Location
Chr.2: 116397154 - 116414788 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006232232.2 XP_006232294.1 5 - 6 2387 133
XM_006232234.2 XP_006232296.1 5 - 6 2257 133

View Full Product Details