The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012178.1 | NP_001012178.1 | 6 - 7 | 1429 | 130 |
XM_006232232.2 | XP_006232294.1 | 7 - 8 | 2557 | 130 | |
XM_006232233.2 | XP_006232295.1 | 6 - 7 | 2474 | 130 | |
XM_006232234.2 | XP_006232296.1 | 7 - 8 | 2427 | 130 | |
GenBank mRNA | BC089853.1 | - | 6 - 7 | 1429 | 130 |