Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sec62
Entrez Gene ID
294912
Gene Name
SEC62 homolog (S. cerevisiae)
Gene Aliases
Ab2-292, Tloc1
Chromosome Location
Chr.2: 116188722 - 116225510 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034129.1 NP_001029301.1 8 - 9 1120 170
XM_006232224.2 XP_006232286.1 4 - 5 702 170
GenBank mRNA AY325207.1 - 8 - 9 1120 170

View Full Product Details