Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Samd7
Entrez Gene ID
310257
Gene Name
sterile alpha motif domain containing 7
Gene Aliases
-
Chromosome Location
Chr.2: 116243243 - 116262612 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191703.1 NP_001178632.1 2 - 3 222 80
XM_006232225.2 XP_006232287.1 3 - 4 746 80

View Full Product Details