Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hnf4g
Entrez Gene ID
365744
Gene Name
hepatocyte nuclear factor 4, gamma
Gene Aliases
-
Chromosome Location
Chr.2: 100225880 - 100372728 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108939.1 NP_001102409.1 6 - 7 847 138
XM_006232165.2 XP_006232227.1 6 - 7 1598 138
XM_006232166.2 XP_006232228.1 7 - 8 1441 138

View Full Product Details