Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc2a2
Entrez Gene ID
25351
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 2
Gene Aliases
GTT2, Glut2
Chromosome Location
Chr.2: 114413431 - 114445418 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012879.2 NP_037011.2 3 - 4 405 87
XM_006232207.2 XP_006232269.1 4 - 5 709 87
GenBank mRNA BC078875.1 - 3 - 4 405 87
J03145.1 - 3 - 4 480 87

View Full Product Details