Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfhx4
Entrez Gene ID
310250
Gene Name
zinc finger homeobox 4
Gene Aliases
RGD1563022
Chromosome Location
Chr.2: 98423569 - 98610396 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191702.1 NP_001178631.1 2 - 3 3025 82

View Full Product Details