Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfhx4
Entrez Gene ID
310250
Gene Name
zinc finger homeobox 4
Gene Aliases
RGD1563022
Chromosome Location
Chr.2: 98423569 - 98610396 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191702.1 NP_001178631.1 9 - 10 4325 70
XM_006232162.2 XP_006232224.1 9 - 10 4431 70
XM_006232164.2 XP_006232226.1 9 - 10 4296 70
XM_008760867.1 XP_008759089.1 9 - 10 4791 70

View Full Product Details