Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Skil
Entrez Gene ID
114208
Gene Name
SKI-like proto-oncogene
Gene Aliases
Skir, Sno
Chromosome Location
Chr.2: 115862932 - 115891309 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006232237.2 XP_006232299.1 5 - 6 2704 85
XM_008760948.1 XP_008759170.1 4 - 5 3710 85
XM_008760949.1 XP_008759171.1 4 - 5 3570 85

View Full Product Details