Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Car1
Entrez Gene ID
310218
Gene Name
carbonic anhydrase I
Gene Aliases
CA-I, Ca1
Chromosome Location
Chr.2: 88185204 - 88227486 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107660.1 NP_001101130.1 2 - 3 125 100
XM_006232111.1 XP_006232173.1 1 - 2 79 100
XM_008760842.1 XP_008759064.1 2 - 3 82 100
GenBank mRNA BC158888.1 - 2 - 3 88 100
FQ225569.1 - 2 - 3 129 100
FQ225719.1 - 2 - 3 119 100
FQ231425.1 - 2 - 3 119 100
FQ233411.1 - 2 - 3 121 100
FQ235246.1 - 2 - 3 129 100

View Full Product Details