The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107660.1 | NP_001101130.1 | 7 - 8 | 753 | 138 |
XM_006232111.1 | XP_006232173.1 | 6 - 7 | 707 | 138 | |
XM_008760842.1 | XP_008759064.1 | 7 - 8 | 710 | 138 | |
GenBank mRNA | BC158888.1 | - | 7 - 8 | 716 | 138 |
FQ225390.1 | - | 5 - 6 | 624 | 138 | |
FQ225569.1 | - | 7 - 8 | 757 | 138 | |
FQ225719.1 | - | 7 - 8 | 748 | 138 | |
FQ231425.1 | - | 7 - 8 | 747 | 138 | |
FQ233411.1 | - | 7 - 8 | 749 | 138 | |
FQ235246.1 | - | 7 - 8 | 757 | 138 |