Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Depdc1b
Entrez Gene ID
310074
Gene Name
DEP domain containing 1B
Gene Aliases
-
Chromosome Location
Chr.2: 39980802 - 40069982 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107651.1 NP_001101121.1 3 - 4 530 66
XM_008760729.1 XP_008758951.1 3 - 4 470 66

View Full Product Details