Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Btf3
Entrez Gene ID
294680
Gene Name
basic transcription factor 3
Gene Aliases
-
Chromosome Location
Chr.2: 28417316 - 28424453 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008309.1 NP_001008310.1 3 - 4 291 114
XM_006231835.2 XP_006231897.1 3 - 4 561 114
GenBank mRNA BC085343.1 - 3 - 4 291 114
FQ220452.1 - 3 - 4 295 114
FQ229173.1 - 3 - 4 297 114

View Full Product Details