The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006231820.2 | XP_006231882.1 | 3 - 4 | 823 | 84 |
GenBank mRNA | BC099754.1 | - | 3 - 4 | 719 | 84 |