Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mtx3
Entrez Gene ID
688905
Gene Name
metaxin 3
Gene Aliases
-
Chromosome Location
Chr.2: 22427452 - 22440813 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003749167.3 XP_003749215.1 5 - 6 542 61
XM_006231788.2 XP_006231850.1 5 - 6 542 61
XM_008760655.1 XP_008758877.1 5 - 6 542 61

View Full Product Details