Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hexb
Entrez Gene ID
294673
Gene Name
hexosaminidase B
Gene Aliases
-
Chromosome Location
Chr.2: 27983925 - 28003260 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011946.1 NP_001011946.1 8 - 9 1169 117
GenBank mRNA BC079376.1 - 8 - 9 1169 117

View Full Product Details