Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mocs2
Entrez Gene ID
294753
Gene Name
molybdenum cofactor synthesis 2
Gene Aliases
-
Chromosome Location
Chr.2: 46980976 - 46992886 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007633.2 NP_001007634.1 2 - 3 277 93
NM_001162413.1 NP_001155885.1 3 - 4 399 93
GenBank mRNA BC079181.1 - 3 - 4 365 93
CO554692.1 - 3 - 4 399 93
FM123325.1 - 2 - 3 231 93
FQ230755.1 - 2 - 3 200 93
FQ235152.1 - 3 - 4 327 93

View Full Product Details