The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014256.2 | NP_001014278.2 | 7 - 8 | 1035 | 79 |
GenBank mRNA | BC079366.1 | - | 7 - 8 | 1035 | 79 |