Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arl15
Entrez Gene ID
689079
Gene Name
ADP-ribosylation factor-like 15
Gene Aliases
Arfrp2
Chromosome Location
Chr.2: 45668852 - 46053167 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008760741.1 XP_008758963.1 1 - 2 162 64

View Full Product Details