Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serf1
Entrez Gene ID
502503
Gene Name
small EDRK-rich factor 1
Gene Aliases
RGD1565105
Chromosome Location
Chr.2: 30345861 - 30352114 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_577983.5 XP_577983.3 2 - 3 409 136

View Full Product Details