Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Thbs4
Entrez Gene ID
29220
Gene Name
thrombospondin 4
Gene Aliases
-
Chromosome Location
Chr.2: 22343727 - 22385855 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017133.1 NP_058829.1 19 - 20 2683 101

View Full Product Details