Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Otp
Entrez Gene ID
294640
Gene Name
orthopedia homeobox
Gene Aliases
-
Chromosome Location
Chr.2: 24546393 - 24554953 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100523.1 NP_001093993.1 1 - 2 183 103

View Full Product Details