Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccbe1
Entrez Gene ID
361341
Gene Name
collagen and calcium binding EGF domains 1
Gene Aliases
RGD1307670
Chromosome Location
Chr.18: 61758754 - 61810116 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772204.1 XP_008770426.1 3 - 4 1237 63

View Full Product Details