Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Exosc8
Entrez Gene ID
295050
Gene Name
exosome component 8
Gene Aliases
-
Chromosome Location
Chr.2: 143925644 - 143932187 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106432.1 NP_001099902.1 1 - 2 505 61
XM_006232332.2 XP_006232394.1 2 - 3 86 61
GenBank mRNA FQ230358.1 - 1 - 2 33 61

View Full Product Details