The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014253.2 | NP_001014275.2 | 5 - 6 | 668 | 76 |
GenBank mRNA | AY995234.1 | - | 5 - 6 | 668 | 76 |
BC086953.1 | - | 5 - 6 | 562 | 76 | |
FQ211813.1 | - | - | 686 | 76 |