Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Id2
Entrez Gene ID
25587
Gene Name
inhibitor of DNA binding 2
Gene Aliases
Ac2-300
Chromosome Location
Chr.6: 44360077 - 44361908 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013060.3 NP_037192.1 2 - 3 480 133
GenBank mRNA BC086391.1 - - 475 137
D10863.1 - 2 - 3 480 133
FQ212969.1 - - 498 138
FQ213471.1 - - 498 137
FQ213662.1 - - 498 136
FQ220574.1 - 2 - 3 498 140
FQ227397.1 - 2 - 3 498 137

View Full Product Details