Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scamp3
Entrez Gene ID
65169
Gene Name
secretory carrier membrane protein 3
Gene Aliases
-
Chromosome Location
Chr.2: 188488904 - 188495147 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006232753.2 XP_006232815.1 2 - 3 711 66

View Full Product Details