Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Schip1
Entrez Gene ID
295105
Gene Name
schwannomin interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.2: 172312290 - 173087236 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100666.1 NP_001094136.1 8 - 9 1487 116
XM_008761079.1 XP_008759301.1 5 - 6 4425 116
XM_008761080.1 XP_008759302.1 5 - 6 1294 116
XM_008761081.1 XP_008759303.1 5 - 6 1170 116
XM_008761082.1 XP_008759304.1 4 - 5 3442 116
XM_008761083.1 XP_008759305.1 4 - 5 741 116
GenBank mRNA BC092618.1 - 4 - 5 459 116
BC169016.1 - 4 - 5 688 116

View Full Product Details