Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pank3
Entrez Gene ID
360511
Gene Name
pantothenate kinase 3
Gene Aliases
-
Chromosome Location
Chr.10: 20591432 - 20615258 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108272.2 NP_001101742.1 3 - 4 698 120

View Full Product Details