The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009624.1 | NP_001009624.1 | 3 - 4 | 309 | 86 |
GenBank mRNA | BC088264.1 | - | 3 - 4 | 309 | 86 |
FQ220730.1 | - | 3 - 4 | 308 | 86 | |
FQ229832.1 | - | 3 - 4 | 308 | 86 |