Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdia2
Entrez Gene ID
287164
Gene Name
protein disulfide isomerase family A, member 2
Gene Aliases
Pdip
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105775.2 NP_001099245.2 - 693 81

View Full Product Details