Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rab26
Entrez Gene ID
171111
Gene Name
RAB26, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.10: 13894271 - 13898578 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133580.1 NP_598264.1 6 - 7 418 74
GenBank mRNA BC061984.1 - 7 - 8 726 74
U18771.1 - 6 - 7 418 74

View Full Product Details