Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krt42
Entrez Gene ID
450231
Gene Name
keratin 42
Gene Aliases
Ka22
Chromosome Location
Chr.10: 88165667 - 88172910 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008816.1 NP_001008816.1 6 - 7 1210 82

View Full Product Details