The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013229.1 | NP_001013247.1 | 1 - 2 | 289 | 90 |
XM_008768087.1 | XP_008766309.1 | 1 - 2 | 285 | 90 | |
GenBank mRNA | BC089885.1 | - | 1 - 2 | 289 | 90 |