The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019325.1 | NP_062198.1 | 19 - 20 | 2392 | 66 |
GenBank mRNA | BC113948.1 | - | 19 - 20 | 2392 | 66 |