The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013116.2 | NP_001013134.2 | 10 - 11 | 1775 | 66 |
XM_008767826.1 | XP_008766048.1 | 10 - 11 | 1913 | 66 | |
GenBank mRNA | BC101901.1 | - | 9 - 10 | 1600 | 66 |