The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031838.3 | NP_114026.3 | 5 - 6 | 811 | 75 |
GenBank mRNA | AI713815.1 | - | 1 - 2 | 75 | |
BC098726.1 | - | 5 - 6 | 800 | 75 | |
FQ210378.1 | - | 6 - 7 | 823 | 75 | |
FQ213061.1 | - | 6 - 7 | 823 | 75 | |
FQ213116.1 | - | 6 - 7 | 820 | 75 | |
FQ214025.1 | - | 6 - 7 | 823 | 75 | |
FQ215811.1 | - | 6 - 7 | 825 | 75 | |
FQ218484.1 | - | 6 - 7 | 824 | 75 | |
FQ218550.1 | - | 6 - 7 | 824 | 75 | |
FQ218641.1 | - | 6 - 7 | 822 | 75 | |
FQ218643.1 | - | 6 - 7 | 826 | 75 | |
FQ219176.1 | - | 6 - 7 | 822 | 75 | |
FQ219844.1 | - | 6 - 7 | 825 | 75 | |
FQ220315.1 | - | 6 - 7 | 823 | 75 | |
FQ220515.1 | - | 6 - 7 | 823 | 75 | |
FQ220831.1 | - | 6 - 7 | 822 | 75 | |
FQ220841.1 | - | 6 - 7 | 823 | 75 | |
FQ220885.1 | - | 6 - 7 | 823 | 75 | |
FQ220902.1 | - | 6 - 7 | 824 | 75 | |
FQ221771.1 | - | 6 - 7 | 823 | 75 | |
FQ222558.1 | - | 6 - 7 | 823 | 75 | |
FQ222607.1 | - | 6 - 7 | 822 | 75 | |
FQ222729.1 | - | 6 - 7 | 823 | 75 | |
FQ222900.1 | - | 6 - 7 | 823 | 75 | |
FQ223242.1 | - | 6 - 7 | 823 | 75 | |
FQ225100.1 | - | 7 - 8 | 696 | 75 | |
FQ225247.1 | - | 6 - 7 | 816 | 75 | |
FQ225937.1 | - | 6 - 7 | 820 | 75 | |
FQ226404.1 | - | 6 - 7 | 823 | 75 | |
FQ226432.1 | - | 6 - 7 | 825 | 75 | |
FQ226962.1 | - | 6 - 7 | 823 | 75 | |
FQ228202.1 | - | 6 - 7 | 823 | 75 | |
FQ229347.1 | - | 6 - 7 | 823 | 75 | |
FQ229368.1 | - | 6 - 7 | 823 | 75 | |
FQ229383.1 | - | 6 - 7 | 821 | 75 | |
FQ229400.1 | - | 6 - 7 | 822 | 75 | |
FQ229467.1 | - | 6 - 7 | 825 | 75 | |
FQ229778.1 | - | 6 - 7 | 823 | 75 | |
FQ230109.1 | - | 6 - 7 | 825 | 75 | |
FQ230697.1 | - | 6 - 7 | 820 | 75 | |
FQ231866.1 | - | 6 - 7 | 823 | 75 | |
FQ233653.1 | - | 6 - 7 | 823 | 75 | |
FQ234367.1 | - | 6 - 7 | 823 | 75 | |
FQ234813.1 | - | 6 - 7 | 823 | 75 | |
FQ235141.1 | - | 6 - 7 | 825 | 75 | |
U92696.1 | - | 6 - 7 | 704 | 75 | |
U92697.1 | - | 6 - 7 | 727 | 75 | |
U92698.1 | - | 6 - 7 | 729 | 75 | |
U92699.1 | - | 6 - 7 | 703 | 75 | |
U92700.1 | - | 6 - 7 | 676 | 75 | |
X57432.1 | - | 6 - 7 | 821 | 75 |