Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd13b
Entrez Gene ID
360575
Gene Name
ankyrin repeat domain 13B
Gene Aliases
RGD1564005
Chromosome Location
Chr.10: 62674406 - 62691949 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008768134.1 XP_008766356.1 6 - 7 909 68
XM_008768135.1 XP_008766357.1 6 - 7 912 68
XM_008768136.1 XP_008766358.1 6 - 7 908 68
XM_008768137.1 XP_008766359.1 6 - 7 908 68
XM_008768138.1 XP_008766360.1 6 - 7 908 68

View Full Product Details