Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd13b
Entrez Gene ID
360575
Gene Name
ankyrin repeat domain 13B
Gene Aliases
RGD1564005
Chromosome Location
Chr.10: 62674406 - 62691949 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008768134.1 XP_008766356.1 8 - 9 1054 134
XM_008768135.1 XP_008766357.1 8 - 9 1057 134
XM_008768136.1 XP_008766358.1 8 - 9 1053 134
XM_008768137.1 XP_008766359.1 8 - 9 1053 134
XM_008768138.1 XP_008766360.1 8 - 9 1053 134

View Full Product Details