Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh3bp5l
Entrez Gene ID
690898
Gene Name
SH3 binding domain protein 5 like
Gene Aliases
RGD1307036
Chromosome Location
Chr.10: 43768601 - 43785702 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127581.1 NP_001121053.1 4 - 5 983 85
XM_008767708.1 XP_008765930.1 5 - 6 1100 85
XM_008767709.1 XP_008765931.1 5 - 6 1070 85
XM_008767710.1 XP_008765932.1 5 - 6 1108 85
GenBank mRNA BC166447.1 - 4 - 5 983 85

View Full Product Details