The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271228.1 | NP_001258157.1 | 9 - 9 | 4333 | 133 |
XM_008773375.1 | XP_008771597.1 | 9 - 9 | 4380 | 133 |