Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pramel
Entrez Gene ID
503464
Gene Name
preferentially expressed antigen in melanoma-like
Gene Aliases
RGD1561307
Chromosome Location
Chr.X: 105937118 - 105939860 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109368.1 NP_001102838.1 2 - 3 884 83

Gene Symbol
LOC100910698
Entrez Gene ID
100910698
Gene Name
leucine-rich repeat-containing protein PRAME-like
Gene Aliases
-
Chromosome Location
Chr.X: 105969570 - 106006755 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003752110.3 XP_003752158.1 7 - 8 1395 83
XM_008773459.1 XP_008771681.1 8 - 9 1323 83

View Full Product Details