Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl4
Entrez Gene ID
317196
Gene Name
kelch-like family member 4
Gene Aliases
RGD1565441
Chromosome Location
Chr.X: 86125917 - 86229565 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108244.1 NP_001101714.1 6 - 7 1316 113

View Full Product Details