Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fndc3c1
Entrez Gene ID
317224
Gene Name
fibronectin type III domain containing 3C1
Gene Aliases
RGD1563846
Chromosome Location
Chr.X: 77416959 - 77559348 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191722.1 NP_001178651.1 - 2827 84

Gene Symbol
LOC103694495
Entrez Gene ID
103694495
Gene Name
fibronectin type III domain containing protein 3C1-like
Gene Aliases
-
Chromosome Location
Chr.X: 77430902 - 77435046 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008773355.1 XP_008771577.1 7 - 8 762 84

View Full Product Details