Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ell2
Entrez Gene ID
309918
Gene Name
elongation factor RNA polymerase II 2
Gene Aliases
-
Chromosome Location
Chr.2: 2456189 - 2525590 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008760603.1 XP_008758825.1 7 - 8 1684 118
XM_008760604.1 XP_008758826.1 7 - 8 970 118

View Full Product Details